accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOClNOO

CAS RN 2516-95-2

IUPAC Name: 5-chloro-2-nitrobenzoic acid
Synonyms: 2-nitro-5-chlorobenzoic acid
Molecular Weight (g/mol): 201.56
Molecular Formula: C7H4ClNO4
InChi Key: ZKUYSJHXBFFGPU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results