accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOFFF

CAS RN 2516-99-6

IUPAC Name: 3,3,3-trifluoropropanoate
Synonyms: 3,3,3-trifluoropropanoate
Molecular Weight (g/mol): 127.04
Molecular Formula: C3H2F3O2
InChi Key: KSNKQSPJFRQSEI-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)