CAS RN 25179-23-1
IUPAC Name:
lithium(1+) 2-methylpropanoate
Synonyms:
lithium(1+) isobutyrate
Molecular Weight (g/mol):
94.04
Molecular Formula:
C4H7LiO2
InChi Key:
IIDVGIFOWJJSIJ-UHFFFAOYSA-M
SMILES:
[Li+].CC(C)C([O-])=O
1
–
1
of
1
results
1
Lithium isobutyrate, 98+%
CAS: 25179-23-1 Molecular Formula: C4H7LiO2 Molecular Weight (g/mol): 94.038 MDL Number: MFCD00210622 InChI Key: IIDVGIFOWJJSIJ-UHFFFAOYSA-M Synonym: lithium isobutyrate,lithium 2-methylpropanoate,acmc-1ccd2,isobutyric acid lithium salt,lithium 1+ isobutyrate,lithium 1+ ion isobutyrate PubChem CID: 23665557 IUPAC Name: lithium;2-methylpropanoate SMILES: [Li+].CC(C)C(=O)[O-]