accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25185-95-9

IUPAC Name: (E)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine
Synonyms: (E)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine
Molecular Weight (g/mol): 190.02
Molecular Formula: C7H5Cl2NO
InChi Key: YBSXDWIAUZOFFV-ONNFQVAWSA-N
SMILES: O\N=C\C1=C(Cl)C=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results