CAS RN 25199-84-2
IUPAC Name:
4-(trifluoromethyl)-1,2-dihydroquinolin-2-one
Synonyms:
4-(trifluoromethyl)-1H-quinolin-2-one
Molecular Weight (g/mol):
213.16
Molecular Formula:
C10H6F3NO
InChi Key:
UUROBWTVZZNDFD-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(=O)NC2=CC=CC=C12
1
–
1
of
1
results
1
4-Trifluoromethyl-2(1H)-quinolinone, 97%, Thermo Scientific Chemicals
CAS: 25199-84-2 Molecular Formula: C10H6F3NO Molecular Weight (g/mol): 213.159 MDL Number: MFCD03407380 InChI Key: UUROBWTVZZNDFD-UHFFFAOYSA-N Synonym: 2-hydroxy-4-trifluoromethyl quinoline,4-trifluoromethyl quinolin-2-ol,4-trifluoromethyl-2 1h-quinolinone,4-trifluoromethyl-1h-quinolin-2-one,2 1h-quinolinone, 4-trifluoromethyl,4-trifluoromethyl quinolin-2 1h-one,pubchem5867,ksc495g1t,4-trifluoromethylquinolin-2-one,4-trifluoromethyl-quinolin-2-ol PubChem CID: 2759347 IUPAC Name: 4-(trifluoromethyl)-1H-quinolin-2-one SMILES: C1=CC=C2C(=C1)C(=CC(=O)N2)C(F)(F)F