accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONHOHONHOO

CAS RN 252049-08-4

IUPAC Name: 4-{[(benzyloxy)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: 4-{[(benzyloxy)methyl]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 460.53
Molecular Formula: C27H28N2O5
InChi Key: KKUPBFMKUPMFRE-UHFFFAOYNA-N
SMILES: OC(=O)C(CCNCOCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)