accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2521-24-6

H2NSCl

CAS RN 2521-24-6

IUPAC Name: 4-chlorobenzene-1-carbothioamide
Synonyms: 4-chlorobenzenecarbothioamide
Molecular Weight (g/mol): 171.64
Molecular Formula: C7H6ClNS
InChi Key: OKPUICCJRDBRJT-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)