accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25217-43-0

IUPAC Name: methyl N-(3,5-dichlorophenyl)carbamate
Synonyms: methyl N-(3,5-dichlorophenyl)carbamate
Molecular Weight (g/mol): 220.05
Molecular Formula: C8H7Cl2NO2
InChi Key: FRSRGACXHCLBTC-UHFFFAOYSA-N
SMILES: COC(=O)NC1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results