accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2

CAS RN 2523-55-9

IUPAC Name: 4-methylcyclohexan-1-amine
Synonyms: 4-methylcyclohexan-1-amine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: KSMVBYPXNKCPAJ-UHFFFAOYSA-N
SMILES: CC1CCC(N)CC1
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)