accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2525-62-4

IUPAC Name: 1-isocyanatohexane
Synonyms: hexyl isocyanate HMI
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: ANJPRQPHZGHVQB-UHFFFAOYSA-N
SMILES: CCCCCCN=C=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results