accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNSOOHH3C

CAS RN 25252-46-4

IUPAC Name: 1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid
Synonyms: 1,3-dimethylthieno[2,3-c]pyrazole-5-carboxylic acid
Molecular Weight (g/mol): 196.22
Molecular Formula: C8H8N2O2S
InChi Key: QRANSYHQSVJLHX-UHFFFAOYSA-N
SMILES: CN1N=C(C)C2=C1SC(=C2)C(O)=O
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)