accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2526-64-9

IUPAC Name: dichlorotriphenyl-λ⁵-phosphane
Synonyms: triphenylphosphine dichloride
Molecular Weight (g/mol): 333.19
Molecular Formula: C18H15Cl2P
InChi Key: ASWXNYNXAOQCCD-UHFFFAOYSA-N
SMILES: ClP(Cl)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results