accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOBr

CAS RN 2527-99-3

IUPAC Name: methyl 5-bromofuran-2-carboxylate
Synonyms: methyl 5-bromofuran-2-carboxylate
Molecular Weight (g/mol): 205.01
Molecular Formula: C6H5BrO3
InChi Key: FBPIDMAELBIRLE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Br)O1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)