accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 253-52-1

IUPAC Name: phthalazine
Synonyms: phthalazine benzopyridazine benzo(d)pyridazine β-phenodiazine
Molecular Weight (g/mol): 130.15
Molecular Formula: C8H6N2
InChi Key: LFSXCDWNBUNEEM-UHFFFAOYSA-N
SMILES: C1=CC=C2C=NN=CC2=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)