accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2530-83-8

IUPAC Name: trimethoxy({3-[(oxiran-2-yl)methoxy]propyl})silane
Synonyms: glycidoxypropyl trimethoxysilane
Molecular Weight (g/mol): 236.34
Molecular Formula: C9H20O5Si
InChi Key: BPSIOYPQMFLKFR-UHFFFAOYNA-N
SMILES: CO[Si](CCCOCC1CO1)(OC)OC
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)