accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25316-40-9

IUPAC Name: hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
Synonyms: hydrogen doxorubicin chloride
Molecular Weight (g/mol): 579.98
Molecular Formula: C27H30ClNO11
InChi Key: MWWSFMDVAYGXBV-FGBJBKNOSA-N
SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (4)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)