accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 253176-93-1

IUPAC Name: tert-butyl 3-(bromomethyl)azetidine-1-carboxylate
Synonyms: tert-butyl 3-(bromomethyl)azetidine-1-carboxylate
Molecular Weight (g/mol): 250.14
Molecular Formula: C9H16BrNO2
InChi Key: PUKCUGDJEPVLPR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(CBr)C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)