accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONI

CAS RN 253176-94-2

IUPAC Name: tert-butyl 3-(iodomethyl)azetidine-1-carboxylate
Synonyms: tert-butyl 3-(iodomethyl)azetidine-1-carboxylate
Molecular Weight (g/mol): 297.14
Molecular Formula: C9H16INO2
InChi Key: CJVGFEARJOREHI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(CI)C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results