accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHClClCl

CAS RN 2533-69-9

IUPAC Name: methyl 2,2,2-trichloroethanimidate
Synonyms: methyl 2,2,2-trichloroethanimidate
Molecular Weight (g/mol): 176.42
Molecular Formula: C3H4Cl3NO
InChi Key: OGBINJLTBZWRRB-UHFFFAOYSA-N
SMILES: COC(=N)C(Cl)(Cl)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results