accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNOOH

CAS RN 253315-06-9

IUPAC Name: 2-(pyrrolidin-1-yl)pyrimidine-5-carboxylic acid
Synonyms: 2-(pyrrolidin-1-yl)pyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 193.21
Molecular Formula: C9H11N3O2
InChi Key: BARCKZNRVNLERF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(N=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results