accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OFBr

CAS RN 253429-19-5

IUPAC Name: 7-bromo-5-fluoro-1-benzofuran
Synonyms: 7-bromo-5-fluoro-1-benzofuran
Molecular Weight (g/mol): 215.02
Molecular Formula: C8H4BrFO
InChi Key: TYBMMMOEVBNTDU-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C2OC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results