CAS RN 25351-24-0
IUPAC Name:
2-[(2,5-dihydroxyphenyl)formamido]acetic acid
Synonyms:
[(2,5-dihydroxyphenyl)formamido]acetic acid
Molecular Weight (g/mol):
211.17
Molecular Formula:
C9H9NO5
InChi Key:
FBFATOOJCPDQOZ-UHFFFAOYSA-N
SMILES:
OC(=O)CNC(=O)C1=CC(O)=CC=C1O
1
–
1
of
1
results
1
Gentisuric Acid, TRC
CAS: 25351-24-0 Molecular Formula: C9H9NO5 Molecular Weight (g/mol): 211.17 Synonym: Gentisuric Acid IUPAC Name: 2-[(2,5-dihydroxybenzoyl)amino]acetic acid SMILES: OC(=O)CNC(=O)c1cc(O)ccc1O