accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25365-71-3

OHN

CAS RN 25365-71-3

IUPAC Name: 2-phenyl-1H-indole-3-carbaldehyde
Synonyms: 2-phenyl-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 221.26
Molecular Formula: C15H11NO
InChi Key: IFIFXODAHZPTEY-UHFFFAOYSA-N
SMILES: O=CC1=C(NC2=CC=CC=C12)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results