accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25366-22-7

CH3HNNNSFFF

CAS RN 25366-22-7

IUPAC Name: N-methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
Synonyms: N-methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 183.15
Molecular Formula: C4H4F3N3S
InChi Key: ARAFQFMXQDBEPU-UHFFFAOYSA-N
SMILES: CNC1=NN=C(S1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results