CAS RN 25369-78-2
IUPAC Name:
5-chloro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms:
5-chloro-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol):
184.64
Molecular Formula:
C7H5ClN2S
InChi Key:
ZZIHEYOZBRPWMB-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2NC(=S)NC2=C1
1
–
1
of
1
results
1
5-Chloro-2-mercaptobenzimidazole, 98%
CAS: 25369-78-2 Molecular Formula: C7H5ClN2S Molecular Weight (g/mol): 184.641 MDL Number: MFCD01658762 InChI Key: ZZIHEYOZBRPWMB-UHFFFAOYSA-N Synonym: 5-chloro-2-mercaptobenzimidazole,6-chloro-1h-benzimidazole-2-thiol,5-chloro-2-benzimidazolethiol,2-benzimidazolinethione, 5-chloro,5-chloro-1,3-dihydro-2h-benzimidazole-2-thione,5-chloro-1h-benzimidazole-2-thiol,2-benzimidazolethiol, 5-chloro,5-chloro-2 3-benzimidazolethione,6-chloro-1h-benzoimidazole-2-thiol,6-chloro-1h-benzo d imidazole-2-thiol PubChem CID: 2056429 IUPAC Name: 5-chloro-1,3-dihydrobenzimidazole-2-thione SMILES: C1=CC2=C(C=C1Cl)NC(=S)N2