accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCH3CH3H3COClOOO

CAS RN 2537-36-2

IUPAC Name: tetramethylazanium perchlorate
Synonyms: tetramethylammonium perchlorate
Molecular Weight (g/mol): 173.59
Molecular Formula: C4H12ClNO4
InChi Key: ZCWKIFAQRXNZCH-UHFFFAOYSA-M
SMILES: C[N+](C)(C)C.[O-][Cl](=O)(=O)=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results