accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OPONOH3C

CAS RN 2537-48-6

IUPAC Name: diethyl (cyanomethyl)phosphonate
Synonyms: diethyl cyanomethylphosphonate
Molecular Weight (g/mol): 177.14
Molecular Formula: C6H12NO3P
InChi Key: KWMBADTWRIGGGG-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC#N)OCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)