CAS RN 25389-94-0
Kanamycin Sulfate (White Powder), Fisher BioReagents
For selecting transformed cells containing kanamycin resistance gene | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol
Kanamycin monosulfate, Cell Culture Grade
CAS: 25389-94-0 Molecular Formula: C18H38N4O15S Molecular Weight (g/mol): 582.575 MDL Number: MFCD00070253 InChI Key: OOYGSFOGFJDDHP-IZQIRFRQSA-N Synonym: Kanamycin A PubChem CID: 134129479 IUPAC Name: (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid SMILES: C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
Thermo Scientific Chemicals Kanamycin sulfate
CAS: 25389-94-0 Molecular Formula: C18H38N4O15S Molecular Weight (g/mol): 582.575 MDL Number: MFCD00070253 InChI Key: OOYGSFOGFJDDHP-IZQIRFRQSA-N PubChem CID: 134129479 IUPAC Name: (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid SMILES: C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
Kanamycin Sulfate, Ultrapure
CAS: 25389-94-0 Molecular Formula: C18H38N4O15S Molecular Weight (g/mol): 582.575 MDL Number: MFCD00070253 InChI Key: OOYGSFOGFJDDHP-IZQIRFRQSA-N Synonym: Kanamycin A PubChem CID: 134129479 IUPAC Name: (2 R,3 S,4 S,5 S,6 R)-2-(aminomethyl)-6-[(1 R,2 S,3 S,4 R,6 S)-4,6-diamino-3-[(2 S,3 R,4 S,5 R,6 R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid SMILES: C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
Kanamycin monosulfate
CAS: 25389-94-0 Molecular Formula: C18H38N4O15S Molecular Weight (g/mol): 582.575 MDL Number: MFCD00070253 InChI Key: OOYGSFOGFJDDHP-IZQIRFRQSA-N Synonym: Kanamycin A PubChem CID: 134129479 IUPAC Name: (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid SMILES: C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
Kanamycin sulfate from Streptomyces kanamycetikus, MP Biomedicals™
CAS: 25389-94-0 Molecular Formula: C18H38N4O15S Molecular Weight (g/mol): 582.58 MDL Number: MFCD00070253 InChI Key: OOYGSFOGFJDDHP-KMCOLRRFSA-N Synonym: Kanamycin A,Kanamycin sulfate salt
Kanamycin monosulfate, MP Biomedicals™
CAS: 25389-94-0 Molecular Formula: C18H38N4O15S Molecular Weight (g/mol): 582.575 InChI Key: OOYGSFOGFJDDHP-IZQIRFRQSA-N Synonym: Kanamycin sulfate salt,Kanamycin A PubChem CID: 134129479 IUPAC Name: (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid SMILES: C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
Melford Kanamycin A
Kanamycin A is an aminoglycoside antibiotic that is widely used in the field of life sciences.
Kanamycin sulfate 100 μg/mL in Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Kanamycin A Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Kanamycin A Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TOKU-E™ Kanamycin ReadyMade™ Solution
Kanamycin ReadyMade™ Solution is provided as a sterile solution of Kanamycin Sulfate, USP formulated in water.