accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25443-63-4

ClHRuHClClHClHHClHCl

CAS RN 25443-63-4

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 319.82
Molecular Formula: Cl6H6Ru
InChi Key: DWXPMVIKISPUHC-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.Cl.Cl.[Ru]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results