accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SNN

CAS RN 25445-77-6

IUPAC Name: 4-phenyl-1,2,3-thiadiazole
Synonyms: 4-phenyl-1,2,3-thiadiazole
Molecular Weight (g/mol): 162.21
Molecular Formula: C8H6N2S
InChi Key: AWPNFXRMNNPKDW-UHFFFAOYSA-N
SMILES: S1C=C(N=N1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results