accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2547-66-2

IUPAC Name: 1,3,5-tribenzyl-1,3,5-triazinane
Synonyms: 1,3,5-tribenzyl-1,3,5-triazinane
Molecular Weight (g/mol): 357.5
Molecular Formula: C24H27N3
InChi Key: VWVZIRPJPFJGFE-UHFFFAOYSA-N
SMILES: C(N1CN(CC2=CC=CC=C2)CN(CC2=CC=CC=C2)C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results