accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NClSOOCl

CAS RN 254749-11-6

IUPAC Name: 2-chloro-4-cyanobenzene-1-sulfonyl chloride
Synonyms: 2-chloro-4-cyanobenzenesulfonyl chloride
Molecular Weight (g/mol): 236.07
Molecular Formula: C7H3Cl2NO2S
InChi Key: GNJOWOAPHOFJBW-UHFFFAOYSA-N
SMILES: ClC1=C(C=CC(=C1)C#N)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results