accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NN

CAS RN 25475-67-6

IUPAC Name: isoquinolin-3-amine
Synonyms: isoquinolin-3-amine
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: VYCKDIRCVDCQAE-UHFFFAOYSA-N
SMILES: NC1=NC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results