accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBHOHOO

CAS RN 25487-66-5

IUPAC Name: [3-(formyloxy)phenyl]boronic acid
Synonyms: 3-(formyloxy)phenylboronic acid
Molecular Weight (g/mol): 165.94
Molecular Formula: C7H7BO4
InChi Key: CGKZLXUMUJKVPS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(OC=O)=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)