accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2549-93-1

H3NNH3

CAS RN 2549-93-1

IUPAC Name: [4-(azaniumylmethyl)cyclohexyl]methanaminium
Synonyms: [4-(ammoniomethyl)cyclohexyl]methanaminium
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: OXIKYYJDTWKERT-UHFFFAOYSA-P
SMILES: [NH3+]CC1CCC(C[NH3+])CC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results