accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO

CAS RN 2550-26-7

IUPAC Name: 4-phenylbutan-2-one
Synonyms: benzylacetone
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: AKGGYBADQZYZPD-UHFFFAOYSA-N
SMILES: CC(=O)CCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (3)
  • (3)