accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Br

CAS RN 2550-36-9

IUPAC Name: (bromomethyl)cyclohexane
Synonyms: (bromomethyl)cyclohexane
Molecular Weight (g/mol): 177.09
Molecular Formula: C7H13Br
InChi Key: UUWSLBWDFJMSFP-UHFFFAOYSA-N
SMILES: BrCC1CCCCC1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)