accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOBr

CAS RN 25542-62-5

IUPAC Name: ethyl 6-bromohexanoate
Synonyms: ethyl 6-bromohexanoate
Molecular Weight (g/mol): 223.11
Molecular Formula: C8H15BrO2
InChi Key: DXBULVYHTICWKT-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCBr
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)