accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FS

CAS RN 2557-78-0

IUPAC Name: (2-fluorophenyl)sulfanide
Synonyms: (2-fluorophenyl)sulfanide
Molecular Weight (g/mol): 127.16
Molecular Formula: C6H4FS
InChi Key: WJTZZPVVTSDNJJ-UHFFFAOYSA-M
SMILES: FC1=CC=CC=C1[S-]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results