accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH3COONHNHCH3CH3

CAS RN 25606-41-1

IUPAC Name: propyl N-[3-(dimethylazaniumyl)propyl]carbamate chloride
Synonyms: propamocarb hcl
Molecular Weight (g/mol): 224.73
Molecular Formula: C9H21ClN2O2
InChi Key: MKIMSXGUTQTKJU-UHFFFAOYSA-N
SMILES: [Cl-].CCCOC(=O)NCCC[NH+](C)C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results