CAS RN 2563-07-7
CAS RN 2563-07-7
IUPAC Name:
2-ethoxy-4-methylphenol
Synonyms:
ethoxy-P-cresol
Molecular Weight (g/mol):
152.19
Molecular Formula:
C9H12O2
InChi Key:
ZWQBCCVEOIYNDL-UHFFFAOYSA-N
SMILES:
CCOC1=CC(C)=CC=C1O