accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 256353-38-5

CH3OOSFFFNH2

CAS RN 256353-38-5

IUPAC Name: methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Synonyms: methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 301.28
Molecular Formula: C13H10F3NO2S
InChi Key: FYUVISRLFPIWPP-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C(=C(S1)C(F)(F)F)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results