accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25637-18-7

IUPAC Name: 4-iodooxane
Synonyms: 4-iodooxane
Molecular Weight (g/mol): 212.03
Molecular Formula: C5H9IO
InChi Key: JTRNQTFTRDPITG-UHFFFAOYSA-N
SMILES: IC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results