accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 256373-91-8

IUPAC Name: ethyl 5-(chlorosulfonyl)furan-3-carboxylate
Synonyms: ethyl 5-(chlorosulfonyl)furan-3-carboxylate
Molecular Weight (g/mol): 238.64
Molecular Formula: C7H7ClO5S
InChi Key: LEKOVVUPHAKDKN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=COC(=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results