CAS RN 256376-24-6
IUPAC Name:
5-cyclopropyl-2-{1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl}pyrimidin-4-amine
Synonyms:
5-cyclopropyl-2-{1-[(2-fluorophenyl)methyl]pyrazolo[3,4-b]pyridin-3-yl}pyrimidin-4-amine
Molecular Weight (g/mol):
360.4
Molecular Formula:
C20H17FN6
InChi Key:
ATOAHNRJAXSBOR-UHFFFAOYSA-N
SMILES:
NC1=NC(=NC=C1C1CC1)C1=NN(CC2=CC=CC=C2F)C2=NC=CC=C12
1
–
2
of
2
results
1
R&D Systems™ BAY 41-2272
CAS: 256376-24-6 Molecular Formula: C20H17FN6
2
BAY 41-2272, TRC
CAS: 256376-24-6 Molecular Formula: C20H17FN6 Molecular Weight (g/mol): 360.39 Synonym: BAY 41-2272,5-Cyclopropyl-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-4-pyrimidinamine IUPAC Name: 5-cyclopropyl-2-[1-[(2-fluorophenyl)methyl]pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-4-amine SMILES: Nc1nc(ncc1C2CC2)c3nn(Cc4ccccc4F)c5ncccc35