accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2564-83-2

CH3H3CH3CH3CNO

CAS RN 2564-83-2

IUPAC Name: (2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Synonyms: tanan tanane tetramethylpiperidine nitroxide oxoammonium tpo tempo tetramethylpiperidinooxy
Molecular Weight (g/mol): 156.25
Molecular Formula: C9H18NO
InChi Key: QYTDEUPAUMOIOP-UHFFFAOYSA-N
SMILES: CC1(C)CCCC(C)(C)N1[O]
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)