accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 256427-77-7

IUPAC Name: 4-phenyl-5-(trifluoromethyl)thiophen-3-amine
Synonyms: 4-phenyl-5-(trifluoromethyl)thiophen-3-amine
Molecular Weight (g/mol): 243.25
Molecular Formula: C11H8F3NS
InChi Key: KBIPYEXWOYDDDL-UHFFFAOYSA-N
SMILES: NC1=CSC(=C1C1=CC=CC=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results