accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25655-41-8

IUPAC Name: 1-ethenylpyrrolidin-2-one; diiodine
Synonyms: povidone-iodine povidone iodine 聚维酮碘
Molecular Weight (g/mol): 364.95
Molecular Formula: C6H9I2NO
InChi Key: CPKVUHPKYQGHMW-UHFFFAOYSA-N
SMILES: II.C=CN1CCCC1=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)