accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3COBOCH3H3C

CAS RN 256652-04-7

IUPAC Name: 4,4,5,5-tetramethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 254.14
Molecular Formula: C16H19BO2
InChi Key: SPPZBAGKKBHZRW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)