accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25676-75-9

IUPAC Name: 4-bromo-1-methyl-1H-imidazole
Synonyms: 4-bromo-1-methylimidazole
Molecular Weight (g/mol): 161
Molecular Formula: C4H5BrN2
InChi Key: IOTSLMMLLXTNNH-UHFFFAOYSA-N
SMILES: CN1C=NC(Br)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results